Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005978.3 | NP_005969.1 | 3 | 855 | ||
GenBank mRNA | BQ953250.1 | - | 550 |