Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286206.1 | NP_001273135.1 | 5 | 408 | ||
NM_014895.3 | NP_055710.2 | 5 | 462 | |||
XM_005248674.3 | XP_005248731.1 | 4 | 585 | |||
XM_005248678.3 | XP_005248735.1 | 5 | 716 | |||
XM_006715380.2 | XP_006715443.1 | 6 | 556 | |||
XM_011535594.2 | XP_011533896.1 | 5 | 716 | |||
XM_017010483.1 | XP_016865972.1 | 4 | 1141 | |||
XM_017010484.1 | XP_016865973.1 | 6 | 759 | |||
GenBank mRNA | AB023226.1 | - | 5 | 369 | ||
AK308190.1 | - | 5 | 429 | |||
BC063632.1 | - | 5 | 435 | |||
DC424230.1 | - | 5 | 444 |