Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005980.2 | NP_005971.1 | 1 | 137 | ||
GenBank mRNA | AF539739.1 | - | 1 | 907 | ||
BC006819.1 | - | 1 | 130 | 3448490 | ||
BI259919.1 | - | 1 | 149 | |||
BT007289.1 | - | 1 | 43 | |||
X65614.1 | - | 1 | 64 |