Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271851.1 | NP_001258780.1 | 5, 6 | 973 | ||
NM_022157.3 | NP_071440.1 | 5, 6 | 1075 | |||
GenBank mRNA | AF272035.1 | - | 5, 6 | 903 | ||
AF323609.1 | - | 5, 6 | 898 | |||
AK023373.1 | - | 1, 2 | 103 | |||
AK298573.1 | - | 4, 5 | 598 | |||
AK298788.1 | - | 5, 6 | 869 | |||
AK315791.1 | - | 5, 6 | 1000 | |||
BC016668.1 | - | 5, 6 | 920 | 4342448 |