Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037633.1 | NP_001032722.1 | 5 | 516 | ||
NM_022464.4 | NP_071909.1 | 4 | 428 | |||
XM_011543570.2 | XP_011541872.1 | 6 | 553 | |||
GenBank mRNA | AF547994.1 | - | 4 | 347 | ||
AJ299442.1 | - | 4 | 406 | |||
AK074624.1 | - | 4 | 420 | |||
AK075177.1 | - | 4 | 357 | |||
AY358950.1 | - | 4 | 374 | |||
BC011568.1 | - | 4 | 396 | 4640182 | ||
BI561751.1 | - | 4 | 459 | |||
CB125000.1 | - | 4 | 428 | |||
KU178743.1 | - | 3 | 310 | |||
KU178744.1 | - | 3 | 310 |