Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022788.4 | NP_073625.1 | 3 | 894 | ||
NM_176876.2 | NP_795345.1 | 2 | 868 | |||
XM_017007069.1 | XP_016862558.1 | 3 | 1152 | |||
GenBank mRNA | AB052684.1 | - | 1 | 594 | ||
AF313449.1 | - | 2 | 668 | |||
AF321815.1 | - | 2 | 665 | |||
AJ320495.1 | - | 1 | 611 | |||
AK292096.1 | - | 3 | 833 | |||
AK303714.1 | - | 4 | 544 | |||
AK316575.1 | - | 3 | 836 | |||
AY136754.1 | - | 1 | 594 | |||
BC017898.1 | - | 3 | 838 | 4251263 |
Set Membership: |
G-protein coupled receptor |