Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000338.2 | NP_000329.2 | 19 | 2555 | ||
NM_001184832.1 | NP_001171761.1 | 19 | 2555 | |||
XM_005254606.1 | XP_005254663.1 | 19 | 2555 | |||
GenBank mRNA | AK295375.1 | - | 18 | 2009 | ||
AK298275.1 | - | 14 | 1775 | |||
AK298312.1 | - | 19 | 2554 | |||
BX647067.1 | - | 14 | 6479 | |||
BX647484.1 | - | 15 | 6462 | |||
EF559316.2 | - | 18 | 2353 | |||
U58130.1 | - | 18 | 2358 |