Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003068.4 | NP_003059.1 | 3 | 899 | ||
GenBank mRNA | AK223368.1 | - | 3 | 888 | ||
AK312661.1 | - | 3 | 888 | |||
BC014890.1 | - | 3 | 876 | 3908245 | ||
BC015895.1 | - | 3 | 856 | 3911047 |