Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278458.1 | NP_001265387.1 | 15 | 3066 | ||
NM_001278460.1 | NP_001265389.1 | 12 | 2889 | |||
NM_001278461.1 | NP_001265390.1 | 11 | 2849 | |||
NM_014900.4 | NP_055715.3 | 12 | 2963 | |||
GenBank mRNA | AB023194.1 | - | 12 | 2957 | ||
AK001813.1 | - | 1 | 28 | |||
AK002057.1 | - | 1 | 75 | |||
AK075181.1 | - | 1 | 701 | |||
AK225849.1 | - | 12 | 2747 | |||
AK294937.1 | - | 15 | 3066 | |||
AL049939.1 | - | 1 | 323 | |||
BC071588.1 | - | 11 | 2851 | 30345650 | ||
BX537877.1 | - | 7 | 4451 | |||
BX648994.1 | - | 1 | 3535 | |||
BX649112.1 | - | 6 | 1832 |