Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021021.3 | NP_066301.1 | 4 | 1283 | ||
XM_011517239.1 | XP_011515541.1 | 4 | 1260 | |||
GenBank mRNA | AF028828.1 | - | 4 | 777 | ||
AK026095.1 | - | 6 | 1426 | |||
AK292655.1 | - | 4 | 1357 | |||
BC098573.1 | - | 4 | 1243 | 30925462 |