Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278458.1 | NP_001265387.1 | 15 | 2443 | ||
NM_001278460.1 | NP_001265389.1 | 12 | 2266 | |||
NM_001278461.1 | NP_001265390.1 | 11 | 2226 | |||
NM_014900.4 | NP_055715.3 | 12 | 2340 | |||
GenBank mRNA | AB023194.1 | - | 12 | 2334 | ||
AK075181.1 | - | 1 | 78 | |||
AK225849.1 | - | 12 | 2123 | |||
AK294937.1 | - | 15 | 2443 | |||
BC071588.1 | - | 11 | 2228 | 30345650 | ||
BX537877.1 | - | 7 | 3828 | |||
BX648994.1 | - | 1 | 2912 | |||
BX649112.1 | - | 6 | 1209 |