Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278458.1 | NP_001265387.1 | 16 | 3632 | ||
NM_001278460.1 | NP_001265389.1 | 13 | 3455 | |||
NM_001278461.1 | NP_001265390.1 | 12 | 3415 | |||
NM_014900.4 | NP_055715.3 | 13 | 3529 | |||
GenBank mRNA | AB023194.1 | - | 13 | 3523 | ||
AK001813.1 | - | 2 | 594 | |||
AK002057.1 | - | 2 | 641 | |||
AK075181.1 | - | 2 | 1267 | |||
AK225849.1 | - | 13 | 3313 | |||
AK294937.1 | - | 16 | 3632 | |||
AL049939.1 | - | 2 | 889 | |||
BC071588.1 | - | 12 | 3417 | 30345650 | ||
BX537877.1 | - | 8 | 5017 | |||
BX648994.1 | - | 2 | 4101 | |||
BX649112.1 | - | 7 | 2398 |