Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007294.3 | NP_009225.1 | 18, 19 | 5427 | ||
NM_007297.3 | NP_009228.2 | 17, 18 | 5335 | |||
NM_007298.3 | NP_009229.2 | 17, 18 | 1902 | |||
NM_007299.3 | NP_009230.2 | 18, 19 | 2077 | |||
NM_007300.3 | NP_009231.2 | 19, 20 | 5490 | |||
NR_027676.1 | 18, 19 | 5331 | ||||
GenBank mRNA | AF005068.1 | - | 18, 19 | 5296 | ||
AY751490.1 | - | 17, 18 | 5195 | |||
BC072418.1 | - | 18, 19 | 2077 | |||
BC115037.1 | - | 19, 20 | 5395 | 7961446 | ||
DQ333386.1 | - | 3, 4 | 269 | |||
DQ363751.1 | - | 6, 7 | 770 | |||
JN686490.1 | - | 17, 18 | 5195 | |||
U14680.1 | - | 18, 19 | 5314 | |||
U64805.1 | - | 17, 18 | 1883 | |||
U68041.1 | - | 4, 5 | 305 |