Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT4H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003168.2 | NP_003159.1 | 3, 4 | 333 | ||
NR_073470.1 | 3, 4 | 335 | ||||
GenBank mRNA | AK129758.1 | - | 3, 4 | 304 | ||
AK311986.1 | - | 3, 4 | 246 | |||
BC002802.2 | - | 3, 4 | 291 | 3636217 | ||
BP196741.1 | - | 3, 4 | 333 | |||
BP318625.1 | - | 3, 4 | 296 | |||
BX331655.2 | - | 3, 4 | 297 | |||
CR407663.1 | - | 3, 4 | 230 | |||
U38817.1 | - | 3, 4 | 272 | |||
U38818.1 | - | 3, 4 | 291 | |||
U43923.1 | - | 3, 4 | 297 |