Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003198.2 | NP_003189.2 | 8 | 1950 | ||
GenBank mRNA | AF116729.1 | - | 8 | 1799 | ||
AK310229.1 | - | 8 | 1936 | |||
AK313079.1 | - | 8 | 1822 | |||
BC002883.2 | - | 8 | 1834 | 3941728 | ||
BC020448.1 | - | 5 | 562 | |||
L47345.1 | - | 8 | 1844 |