Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000459.4 | NP_000450.2 | 12 | 2114 | ||
NM_001290077.1 | NP_001277006.1 | 11 | 1985 | |||
NM_001290078.1 | NP_001277007.1 | 10 | 1673 | |||
XM_005251561.1 | XP_005251618.1 | 12 | 1781 | |||
XM_005251563.1 | XP_005251620.1 | 11 | 1652 | |||
GenBank mRNA | AB086825.1 | - | 11 | 1641 | ||
AB208796.1 | - | 12 | 1792 | |||
AK291775.1 | - | 12 | 1802 | |||
AK294887.1 | - | 10 | 1674 | |||
AK295043.1 | - | 11 | 1711 | |||
AK308447.1 | - | 9 | 1225 | |||
BC035514.1 | - | 12 | 1789 | 5228999 | ||
L06139.1 | - | 12 | 1812 |