Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003247.3 | NP_003238.2 | 7 | 1303 | ||
GenBank mRNA | AK292429.1 | - | 7 | 1260 | ||
BC146676.1 | - | 6 | 1110 | 40148868 | ||
BC150175.1 | - | 6 | 1114 | 40147958 | ||
L12350.1 | - | 7 | 1222 |