Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253357.1 | NP_001240286.1 | 18 | 3050 | ||
NM_005424.4 | NP_005415.1 | 18 | 3133 | |||
XM_005271163.1 | XP_005271220.1 | 17 | 2947 | |||
XM_006710869.1 | XP_006710932.1 | 18 | 3267 | |||
GenBank mRNA | AF217976.1 | - | 8 | 1217 | ||
AK225560.1 | - | 18 | 2792 | |||
AK296735.1 | - | 13 | 2109 | |||
AK300399.1 | - | 18 | 2940 | |||
AL833389.1 | - | 3 | 1585 | |||
BC038239.1 | - | 18 | 3035 | 5767075 | ||
X60957.1 | - | 18 | 2992 |