Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032283.2 | NP_001027454.1 | 1 | 337 | ||
NM_001032284.2 | NP_001027455.1 | 1 | 337 | |||
NM_001307975.1 | NP_001294904.1 | 1 | 337 | |||
NM_003276.2 | NP_003267.1 | 1 | 337 | |||
XM_005269132.3 | XP_005269189.1 | 1 | 328 | |||
GenBank mRNA | AB209297.1 | - | 1 | 274 | ||
AF070631.1 | - | 1 | 278 | |||
AK315221.1 | - | 1 | 266 | |||
BC053675.1 | - | 1 | 283 | 6170639 | ||
DB239672.1 | - | 1 | 337 | |||
EF028063.1 | - | 1 | 58 | |||
S76736.1 | - | 1 | 242 | |||
U09086.1 | - | 1 | 246 | |||
U09087.1 | - | 1 | 279 | |||
U09088.1 | - | 1 | 279 |