Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022170.1 | NP_071496.1 | 7 | 638 | ||
NM_031992.1 | NP_114381.1 | 6 | 578 | |||
XM_011516556.1 | XP_011514858.1 | 7 | 671 | |||
XM_011516557.1 | XP_011514859.1 | 6 | 596 | |||
GenBank mRNA | AK290676.1 | - | 6 | 598 | ||
AK314749.1 | - | 6 | 680 | |||
BC010021.1 | - | 1 | 576 | 2960402 | ||
BC021214.2 | - | 6 | 576 | 4139578 | ||
BC066928.1 | - | 1 | 595 | 5263716 | ||
D26068.1 | - | 6 | 578 |