Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297588.1 | NP_001284517.1 | 5 | 496 | ||
NM_001297589.1 | NP_001284518.1 | 5 | 506 | |||
NM_013359.2 | NP_037491.2 | 6 | 644 | |||
XM_017027232.1 | XP_016882721.1 | 5 | 523 | |||
XM_017027233.1 | XP_016882722.1 | 4 | 440 | |||
GenBank mRNA | AF187987.1 | - | 6 | 644 | ||
AK314207.1 | - | 5 | 491 | |||
BC111981.1 | - | 5 | 399 | 8327449 | ||
BC113426.1 | - | 5 | 399 | 8322478 | ||
BC143659.1 | - | 6 | 547 | |||
BP337374.1 | - | 5 | 507 | |||
DA493401.1 | - | 5 | 497 |