Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134655.1 | NP_001128127.1 | 2 | 544 | ||
NM_004220.2 | NP_004211.1 | 2 | 563 | |||
NR_104432.1 | 2 | 563 | ||||
XM_011522652.2 | XP_011520954.1 | 2 | 531 | |||
GenBank mRNA | AF017433.1 | - | 3 | 486 | ||
AK055337.1 | - | 2 | 460 | |||
AK289834.1 | - | 2 | 512 | |||
AK291789.1 | - | 2 | 476 | |||
BC007287.1 | - | 2 | 481 | 3347511 | ||
DA115503.1 | - | 2 | 544 | |||
KU178402.1 | - | 1 | 116 | |||
KU178403.1 | - | 1 | 116 |