Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010972.1 | NP_001010972.1 | 5 | 965 | ||
NM_003461.4 | NP_003452.1 | 5 | 968 | |||
XM_011516569.2 | XP_011514871.2 | 4 | 863 | |||
XM_017012587.1 | XP_016868076.1 | 5 | 1058 | |||
GenBank mRNA | AK130138.1 | - | 1 | 258 | ||
AK298925.1 | - | 3 | 714 | |||
AK299005.1 | - | 4 | 684 | |||
AK302309.1 | - | 4 | 776 | |||
AK316227.1 | - | 4 | 660 | |||
BC002323.2 | - | 5 | 837 | |||
BC008743.2 | - | 5 | 932 | 3345843 | ||
BC009360.2 | - | 5 | 877 | 4301542 | ||
BC010031.2 | - | 5 | 865 | 3506023 | ||
BC017183.2 | - | 4 | 494 | |||
CR457431.1 | - | 4 | 810 | |||
U15158.1 | - | 3 | 573 | |||
X94991.1 | - | 4 | 820 | |||
X95735.1 | - | 5 | 881 |