Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024329.5 | NP_077305.2 | 1 | 363 | ||
XM_005246000.3 | XP_005246057.1 | 1 | 361 | |||
GenBank mRNA | BC007233.1 | - | 1 | 321 | 2961350 | |
BC014923.2 | - | 1 | 317 | 3909362 | ||
BC023611.2 | - | 1 | 313 | 4638660 | ||
BC068473.1 | - | 1 | 336 | 5267148 | ||
BI753983.1 | - | 1 | 366 |