Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004057.2 | NP_004048.1 | 2 | 150 | ||
XM_017029841.1 | XP_016885330.1 | 2 | 617 | |||
GenBank mRNA | BC112174.1 | - | 1 | 116 | 8327642 | |
L13220.1 | - | 2 | 153 | |||
X65869.1 | - | 2 | 150 |