Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136040.2 | NP_001129512.1 | 3 | 208 | ||
NM_001142565.1 | NP_001136037.1 | 3 | 208 | |||
NM_024811.3 | NP_079087.3 | 3 | 284 | |||
XM_005274298.4 | XP_005274355.1 | 3 | 294 | |||
XM_005274299.4 | XP_005274356.1 | 3 | 211 | |||
XM_005274303.4 | XP_005274360.1 | 3 | 165 | |||
XM_011545257.2 | XP_011543559.1 | 3 | 294 | |||
XM_011545258.2 | XP_011543560.1 | 3 | 165 | |||
XM_011545259.2 | XP_011543561.1 | 3 | 165 | |||
XM_011545260.2 | XP_011543562.1 | 3 | 208 | |||
XM_011545261.2 | XP_011543563.1 | 3 | 209 | |||
XM_011545262.2 | XP_011543564.1 | 3 | 166 | |||
XM_011545263.2 | XP_011543565.1 | 3 | 165 | |||
XM_017018345.1 | XP_016873834.1 | 3 | 294 | |||
GenBank mRNA | AJ275970.1 | - | 2 | 74 | ||
AK092037.1 | - | 2 | 138 | |||
AK096343.1 | - | 3 | 164 | |||
AK294578.1 | - | 3 | 164 | |||
AK307190.1 | - | 3 | 330 | |||
AL832667.1 | - | 3 | 199 | |||
AL833332.1 | - | 3 | 164 | |||
BC018135.1 | - | 3 | 174 | 3913173 | ||
BX537888.1 | - | 3 | 189 | |||
DA194038.1 | - | 3 | 284 | |||
DA804579.1 | - | 3 | 208 |