Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025245.2 | NP_079521.1 | 5 | 973 | ||
NR_038198.1 | 5 | 872 | ||||
XM_006722911.3 | XP_006722974.1 | 4 | 554 | |||
XM_011528320.2 | XP_011526622.1 | 6 | 1019 | |||
XM_011528322.2 | XP_011526624.1 | 4 | 608 | |||
XM_017027326.1 | XP_016882815.1 | 6 | 654 | |||
XM_017027327.1 | XP_016882816.1 | 6 | 635 | |||
XM_017027328.1 | XP_016882817.1 | 6 | 1019 | |||
XM_017027329.1 | XP_016882818.1 | 3 | 444 | |||
GenBank mRNA | AJ300182.1 | - | 4 | 554 | ||
AK097427.1 | - | 5 | 710 | |||
AK301631.1 | - | 5 | 637 | |||
BC141859.1 | - | 5 | 973 | 8860384 | ||
BC143588.1 | - | 5 | 648 | |||
BC143589.1 | - | 5 | 659 |