Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143957.2 | NP_001137429.1 | 3 | 1510 | ||
NM_030784.3 | NP_110411.1 | 2 | 1392 | |||
XM_006715570.3 | XP_006715633.1 | 2 | 1462 | |||
XM_011536153.2 | XP_011534455.1 | 3 | 1453 | |||
XM_011536154.2 | XP_011534456.1 | 3 | 1326 | |||
XM_011536155.2 | XP_011534457.1 | 3 | 1505 | |||
XM_011536157.2 | XP_011534459.1 | 2 | 1228 | |||
XM_011536159.2 | XP_011534461.1 | 2 | 1397 | |||
XM_017011334.1 | XP_016866823.1 | 2 | 1920 | |||
GenBank mRNA | AB030566.1 | - | 2 | 1239 | ||
AB209347.1 | - | 2 | 1378 | |||
AK289839.1 | - | 2 | 1392 | |||
AK313535.1 | - | 2 | 1361 | |||
BC067465.1 | - | 1 | 1067 | 6971763 | ||
BC067466.1 | - | 1 | 1068 | 6971764 | ||
BC067467.1 | - | 1 | 1068 | 6971765 | ||
BC067468.1 | - | 1 | 1068 | 6971766 | ||
BC067469.1 | - | 1 | 1068 | 6971767 | ||
FJ348260.1 | - | 1 | 1032 |
Set Membership: |
G-protein coupled receptor |