Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282546.1 | NP_001269475.1 | 4 | 503 | ||
NM_001282547.1 | NP_001269476.1 | 4 | 488 | |||
NM_032017.2 | NP_114406.1 | 5 | 725 | |||
GenBank mRNA | AK024504.1 | - | 3 | 287 | ||
AK075048.1 | - | 4 | 485 | |||
AL834137.1 | - | 5 | 682 | |||
AY198395.1 | - | 4 | 458 | |||
BC007835.2 | - | 4 | 456 | 4109652 | ||
BC008344.1 | - | 4 | 464 | 3530483 | ||
DA375448.1 | - | 4 | 488 |