Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016620.3 | NP_057704.2 | 4 | 521 | ||
NM_032186.4 | NP_115562.3 | 4 | 551 | |||
NM_201269.2 | NP_958357.1 | 6 | 4195 | |||
XM_005271257.4 | XP_005271314.1 | 8 | 4654 | |||
XM_005271260.4 | XP_005271317.1 | 7 | 4631 | |||
XM_011542258.2 | XP_011540560.1 | 7 | 4332 | |||
XM_011542259.2 | XP_011540561.1 | 9 | 5008 | |||
XM_011542260.2 | XP_011540562.1 | 9 | 4912 | |||
XM_011542261.2 | XP_011540563.1 | 7 | 4253 | |||
XM_017002487.1 | XP_016857976.1 | 8 | 4443 | |||
XM_017002488.1 | XP_016857977.1 | 7 | 4531 | |||
XM_017002489.1 | XP_016857978.1 | 6 | 4506 | |||
XM_017002490.1 | XP_016857979.1 | 7 | 4318 | |||
XM_017002491.1 | XP_016857980.1 | 6 | 4130 | |||
XM_017002492.1 | XP_016857981.1 | 5 | 1541 | |||
XM_017002493.1 | XP_016857982.1 | 5 | 887 | |||
XM_017002494.1 | XP_016857983.1 | 5 | 664 | |||
GenBank mRNA | AB033047.1 | - | 4 | 3778 | ||
AK001587.1 | - | 4 | 1960 | |||
AK023596.1 | - | 4 | 1134 | |||
AK291520.1 | - | 4 | 321 | |||
BC031003.1 | - | 3 | 582 | |||
BC050656.2 | - | 4 | 470 | |||
BC063683.1 | - | 5 | 3889 | |||
BC110841.1 | - | 4 | 369 | |||
BC132776.1 | - | 6 | 4010 | 40146798 | ||
BC132778.1 | - | 6 | 4010 | 40146800 | ||
BI760689.1 | - | 517 | ||||
DA147194.1 | - | 4 | 523 |