Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160328.1 | NP_001153800.1 | 4 | 1205 | ||
NM_001160329.1 | NP_001153801.1 | 4 | 841 | |||
NM_032298.2 | NP_115674.1 | 2 | 1276 | |||
XM_011527390.2 | XP_011525692.1 | 4 | 2263 | |||
XM_011527391.2 | XP_011525693.1 | 4 | 913 | |||
GenBank mRNA | AL136594.1 | - | 2 | 1276 | ||
BC028379.1 | - | 4 | 1201 | 4819777 | ||
BC031067.1 | - | 4 | 824 | 5275216 |