Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290137.1 | NP_001277066.1 | 7 | 807 | ||
NM_001290139.1 | NP_001277068.1 | 7 | 877 | |||
NM_001323635.1 | NP_001310564.1 | 7 | 912 | |||
NM_001323636.1 | NP_001310565.1 | 7 | 912 | |||
NM_032304.3 | NP_115680.1 | 6 | 838 | |||
XM_005255631.4 | XP_005255688.1 | 7 | 814 | |||
XM_011522711.1 | XP_011521013.1 | 7 | 742 | |||
XM_011522712.1 | XP_011521014.1 | 6 | 1069 | |||
XM_017023773.1 | XP_016879262.1 | 7 | 702 | |||
XM_017023774.1 | XP_016879263.1 | 7 | 702 | |||
GenBank mRNA | AK054841.1 | - | 6 | 821 | ||
AK098340.1 | - | 2 | 1466 | |||
AK293575.1 | - | 6 | 838 | |||
BC004353.1 | - | 6 | 813 | 3351990 | ||
BC015008.1 | - | 6 | 766 | 4862387 | ||
BC033796.1 | - | 7 | 855 | 5173915 | ||
BC036747.1 | - | 7 | 855 | |||
BG821643.1 | - | 7 | 624 | |||
BI333301.1 | - | 6 | 674 | |||
BI834617.1 | - | 2 | 143 | |||
BQ067068.1 | - | 2 | 125 |