Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEGF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256545.1 | NP_001243474.1 | 3 | 493 | ||
NM_001308119.1 | NP_001295048.1 | 4 | 544 | |||
NM_001308121.1 | NP_001295050.1 | 3 | 493 | |||
NM_032446.2 | NP_115822.1 | 4 | 544 | |||
XM_011543694.1 | XP_011541996.1 | 3 | 256 | |||
XM_017009987.1 | XP_016865476.1 | 5 | 789 | |||
GenBank mRNA | AB058676.1 | - | 4 | 385 | ||
AK123568.1 | - | 5 | 659 | |||
AK307919.1 | - | 3 | 412 | |||
BC020198.1 | - | 4 | 461 | |||
BC152478.1 | - | 4 | 385 | 8862616 | ||
CR749437.1 | - | 3 | 427 | |||
DA240513.1 | - | 3 | 493 |