Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282062.1 | NP_001268991.1 | 7 | 837 | ||
NM_001282063.1 | NP_001268992.1 | 7 | 799 | |||
NM_001282068.1 | NP_001268997.1 | 6 | 764 | |||
NM_003586.2 | NP_003577.2 | 6 | 798 | |||
NR_104089.1 | 6 | 581 | ||||
NR_104090.1 | 6 | 589 | ||||
XM_011545975.1 | XP_011544277.1 | 8 | 1223 | |||
XM_017023776.1 | XP_016879265.1 | 10 | 1441 | |||
XM_017023777.1 | XP_016879266.1 | 7 | 1378 | |||
XM_017023778.1 | XP_016879267.1 | 6 | 852 | |||
GenBank mRNA | AK090829.1 | - | 1 | 1455 | ||
AK293651.1 | - | 6 | 538 | |||
AK309158.1 | - | 1 | 34 | |||
BC041769.1 | - | 7 | 800 | 4819032 | ||
BC055284.1 | - | 6 | 798 | 5767656 | ||
BC063436.1 | - | 7 | 763 | 5394616 | ||
BI757951.1 | - | 8 | 708 | |||
BX460259.2 | - | 6 | 510 | |||
D31897.1 | - | 6 | 719 |