Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318005.1 | NP_001304934.1 | 5 | 1906 | ||
NM_032788.2 | NP_116177.1 | 5 | 1526 | |||
XM_006712806.2 | XP_006712869.1 | 5 | 1337 | |||
GenBank mRNA | AK027363.1 | - | 5 | 1459 | ||
AL832263.1 | - | 3 | 3522 | |||
AL833754.1 | - | 1 | 544 | |||
BC074900.2 | - | 4 | 1120 | 30915480 | ||
BC074901.2 | - | 4 | 1120 | 30915251 | ||
BC110524.1 | - | 5 | 1658 | 40034154 | ||
BC110525.1 | - | 5 | 1278 | 40034155 |