Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258452.1 | NP_001245381.1 | 9 | 1606 | ||
NM_001258453.1 | NP_001245382.1 | 10 | 1678 | |||
NM_032807.4 | NP_116196.3 | 10 | 1694 | |||
NM_178150.2 | NP_835363.1 | 9 | 1580 | |||
XM_011519748.2 | XP_011518050.1 | 11 | 2274 | |||
XM_017016842.1 | XP_016872331.1 | 10 | 2821 | |||
XM_017016843.1 | XP_016872332.1 | 11 | 2205 | |||
XM_017016844.1 | XP_016872333.1 | 9 | 1590 | |||
XM_017016845.1 | XP_016872334.1 | 9 | 1947 | |||
XM_017016846.1 | XP_016872335.1 | 8 | 1399 | |||
GenBank mRNA | AF380349.1 | - | 10 | 1694 | ||
AF454502.1 | - | 9 | 1521 | |||
AF456237.1 | - | 9 | 1536 | |||
AK027496.1 | - | 6 | 663 | |||
AK095343.1 | - | 9 | 1554 | |||
AK226128.1 | - | 9 | 2104 | |||
AK315173.1 | - | 8 | 1296 | |||
BC020266.1 | - | 7 | 1088 | |||
BC032674.1 | - | 7 | 1032 | |||
BC080629.1 | - | 7 | 1243 | |||
BC110884.1 | - | 10 | 1603 | 6147373 | ||
BC113375.1 | - | 9 | 1527 | 8322427 | ||
BC113377.1 | - | 9 | 1527 | 8322429 |