Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258452.1 | NP_001245381.1 | 6 | 1334 | ||
NM_001258453.1 | NP_001245382.1 | 7 | 1406 | |||
NM_032807.4 | NP_116196.3 | 7 | 1422 | |||
NM_178150.2 | NP_835363.1 | 6 | 1308 | |||
XM_011519748.2 | XP_011518050.1 | 8 | 2002 | |||
XM_017016842.1 | XP_016872331.1 | 7 | 2549 | |||
XM_017016843.1 | XP_016872332.1 | 8 | 1933 | |||
XM_017016844.1 | XP_016872333.1 | 6 | 1318 | |||
XM_017016845.1 | XP_016872334.1 | 6 | 1675 | |||
XM_017016846.1 | XP_016872335.1 | 5 | 1164 | |||
GenBank mRNA | AF380349.1 | - | 7 | 1422 | ||
AF454502.1 | - | 6 | 1249 | |||
AF456237.1 | - | 6 | 1264 | |||
AK027496.1 | - | 3 | 391 | |||
AK095343.1 | - | 6 | 1282 | |||
AK122753.1 | - | 6 | 1334 | |||
AK226128.1 | - | 6 | 1832 | |||
AK315173.1 | - | 5 | 1024 | |||
BC006430.1 | - | 4 | 999 | |||
BC020266.1 | - | 4 | 816 | |||
BC032674.1 | - | 4 | 760 | |||
BC080629.1 | - | 4 | 971 | |||
BC110884.1 | - | 7 | 1331 | 6147373 | ||
BC113375.1 | - | 6 | 1255 | 8322427 | ||
BC113377.1 | - | 6 | 1255 | 8322429 |