Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA2 SIRNA Assays ›
Gene Symbol:
PPFIA2
Gene Name:
PTPRF interacting protein alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 81257975 - 81759748 on Build GRCh38
UniGene ID:
Hs.506216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224983
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220473.2
NP_001207402.1
19, 20
2685
NM_001220474.2
NP_001207403.1
18, 19
2631
NM_001220475.2
NP_001207404.1
19, 20
2497
NM_001220476.2
NP_001207405.1
19, 20
2497
NM_001220477.2
NP_001207406.1
17, 18
2504
NM_001220478.2
NP_001207407.1
16, 17
2429
NM_001220479.2
NP_001207408.1
10, 11
1246
NM_001220480.2
NP_001207409.1
11, 12
1179
NM_001282536.1
NP_001269465.1
17, 18
2192
NM_003625.4
NP_003616.2
20, 21
2548
XM_006719670.3
XP_006719733.1
17, 18
9740
XM_011538906.2
XP_011537208.1
18, 19
2625
XM_011538907.2
XP_011537209.1
18, 19
2625
XM_017020079.1
XP_016875568.1
20, 21
2532
XM_017020080.1
XP_016875569.1
19, 20
2679
XM_017020081.1
XP_016875570.1
19, 20
2679
XM_017020082.1
XP_016875571.1
20, 21
2532
XM_017020083.1
XP_016875572.1
19, 20
2679
XM_017020084.1
XP_016875573.1
19, 20
2679
XM_017020085.1
XP_016875574.1
19, 20
2679
XM_017020086.1
XP_016875575.1
20, 21
2532
XM_017020087.1
XP_016875576.1
19, 20
2666
XM_017020088.1
XP_016875577.1
19, 20
2912
XM_017020089.1
XP_016875578.1
18, 19
2625
XM_017020090.1
XP_016875579.1
20, 21
2532
XM_017020091.1
XP_016875580.1
19, 20
2679
XM_017020092.1
XP_016875581.1
19, 20
2679
XM_017020093.1
XP_016875582.1
19, 20
2679
XM_017020094.1
XP_016875583.1
19, 20
2457
XM_017020095.1
XP_016875584.1
19, 20
2679
XM_017020096.1
XP_016875585.1
18, 19
2604
XM_017020097.1
XP_016875586.1
20, 21
2532
XM_017020098.1
XP_016875587.1
20, 21
2532
XM_017020099.1
XP_016875588.1
18, 19
2625
XM_017020100.1
XP_016875589.1
19, 20
2679
XM_017020101.1
XP_016875590.1
20, 21
2532
XM_017020102.1
XP_016875591.1
18, 19
2591
XM_017020103.1
XP_016875592.1
19, 20
2679
XM_017020104.1
XP_016875593.1
19, 20
2679
XM_017020105.1
XP_016875594.1
18, 19
2604
XM_017020106.1
XP_016875595.1
19, 20
2679
XM_017020107.1
XP_016875596.1
20, 21
2532
XM_017020108.1
XP_016875597.1
20, 21
2532
XM_017020109.1
XP_016875598.1
19, 20
2679
XM_017020110.1
XP_016875599.1
18, 19
2837
XM_017020111.1
XP_016875600.1
19, 20
2679
XM_017020112.1
XP_016875601.1
17, 18
2504
XM_017020113.1
XP_016875602.1
17, 18
2504
XM_017020114.1
XP_016875603.1
17, 18
2504
XM_017020115.1
XP_016875604.1
17, 18
2504
XM_017020116.1
XP_016875605.1
17, 18
2504
XM_017020117.1
XP_016875606.1
16, 17
2239
XM_017020118.1
XP_016875607.1
18, 19
2479
XM_017020119.1
XP_016875608.1
17, 18
2504
XM_017020120.1
XP_016875609.1
17, 18
2192
GenBank mRNA
AB210009.1
-
19, 20
2683
AF034799.1
-
20, 21
2556
AK123372.1
-
11, 12
1114
AK126971.1
-
18, 19
2591
AK294505.1
-
16, 17
2239
AK296380.1
-
10, 11
1217
AK299853.1
-
17, 18
2453
AK302658.1
-
17, 18
2163
BC104912.1
-
19, 20
2481
8143915
BC143485.1
-
19, 20
2481
9051993
DA368637.1
-
4, 5
502
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
neurotransmitter secretion
glutamate secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-v5ndd:80/100.66.76.213:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline