Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF020833.1 | - | 4 | 229 | ||
AF094850.1 | - | 4 | 247 | |||
AK291320.1 | - | 4 | 248 | |||
AK293374.1 | - | 4 | 332 | |||
AK296377.1 | - | 2 | 878 | |||
AK309944.1 | - | 4 | 246 | |||
AK311624.1 | - | 4 | 381 | |||
AK315896.1 | - | 4 | 253 | |||
AK316040.1 | - | 3 | 377 | |||
AK316335.1 | - | 3 | 383 | |||
BC000733.2 | - | 4 | 248 | 3504640 | ||
BC008469.1 | - | 4 | 249 | 4279770 | ||
BF701069.1 | - | 4 | 266 | |||
BT006889.1 | - | 4 | 206 | |||
CR457134.1 | - | 4 | 206 | |||
DC356593.1 | - | 4 | 271 | |||
U96074.1 | - | 4 | 215 |