Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001756.3 | NP_001747.2 | 5 | 1155 | ||
GenBank mRNA | AK225591.1 | - | 6 | 1065 | ||
AK290821.1 | - | 5 | 1140 | |||
AK313760.1 | - | 5 | 1140 | |||
BC017462.1 | - | 5 | 1109 | |||
BC056259.1 | - | 5 | 1130 | 5810260 | ||
BC058021.1 | - | 5 | 1139 | 6714173 | ||
J02943.1 | - | 4 | 1086 |