Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003817.3 | NP_003808.2 | 12 | 1250 | ||
XM_017013943.1 | XP_016869432.1 | 12 | 1137 | |||
GenBank mRNA | AF090327.1 | - | 7 | 639 | ||
AF215824.1 | - | 12 | 1221 | |||
AK292492.1 | - | 12 | 1254 | |||
BC043207.2 | - | 6 | 583 |