Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206897.1 | NP_001193826.1 | 6 | 879 | ||
NM_003888.3 | NP_003879.2 | 5 | 782 | |||
NM_170696.2 | NP_733797.1 | 5 | 782 | |||
NM_170697.2 | NP_733798.1 | 3 | 386 | |||
GenBank mRNA | AB015226.1 | - | 5 | 552 | ||
AB015227.1 | - | 3 | 315 | |||
AB015228.1 | - | 3 | 277 | |||
AK128709.1 | - | 5 | 1277 | |||
AK294981.1 | - | 5 | 482 | |||
AK303057.1 | - | 6 | 624 | |||
BC030589.2 | - | 5 | 540 | 4826743 | ||
BX443600.2 | - | 3 | 386 |