Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103.3 | NP_001094.1 | 19 | 2547 | ||
NM_001278343.1 | NP_001265272.1 | 19 | 2547 | |||
NM_001278344.1 | NP_001265273.1 | 21 | 2744 | |||
GenBank mRNA | AB209521.1 | - | 21 | 2688 | ||
AK294906.1 | - | 17 | 2058 | |||
AK297469.1 | - | 13 | 1692 | |||
AK297618.1 | - | 12 | 1629 | |||
AK297628.1 | - | 12 | 1407 | |||
AK315250.1 | - | 19 | 2547 | |||
BC047901.2 | - | 19 | 2479 | 6199155 | ||
BC051770.1 | - | 19 | 2492 | 6198688 | ||
M86406.1 | - | 19 | 2500 |