Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033229.2 | NP_150232.2 | 7 | 2104 | ||
XM_011514987.1 | XP_011513289.1 | 8 | 1343 | |||
XM_011514988.2 | XP_011513290.1 | 7 | 1611 | |||
GenBank mRNA | AF220132.1 | - | 7 | 1625 | ||
AF220133.1 | - | 2 | 317 | |||
AK290280.1 | - | 7 | 1810 | |||
BC038585.1 | - | 7 | 1705 | 5758860 | ||
U34249.1 | - | 7 | 2118 |