Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318063.1 | NP_001304992.1 | 5 | 826 | ||
XM_005264633.2 | XP_005264690.1 | 5 | 1557 | |||
XM_005264634.2 | XP_005264691.1 | 4 | 1450 | |||
XM_011533148.2 | XP_011531450.1 | 6 | 2256 | |||
XM_011533149.2 | XP_011531451.1 | 5 | 2149 | |||
XM_011533150.2 | XP_011531452.1 | 7 | 2457 | |||
XM_011533151.2 | XP_011531453.1 | 6 | 1611 | |||
XM_011533152.2 | XP_011531454.1 | 6 | 2328 | |||
XM_011533153.2 | XP_011531455.1 | 5 | 1504 | |||
XM_017005225.1 | XP_016860714.1 | 7 | 1377 | |||
XM_017005226.1 | XP_016860715.1 | 5 | 1629 | |||
GenBank mRNA | AB074759.1 | - | 2 | 487 | ||
AK058055.1 | - | 2 | 260 | |||
AL541274.3 | - | 5 | 826 | |||
BC008981.1 | - | 2 | 487 | 3456620 | ||
BC034808.1 | - | 2 | 1226 | 5167103 |