Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKMYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258451.1 | NP_001245380.1 | 3 | 487 | ||
NM_004203.4 | NP_004194.3 | 3 | 623 | |||
NM_182687.2 | NP_872629.1 | 3 | 623 | |||
XM_011522734.2 | XP_011521036.1 | 3 | 584 | |||
XM_011522735.2 | XP_011521037.1 | 3 | 410 | |||
XM_011522736.2 | XP_011521038.1 | 2 | 417 | |||
GenBank mRNA | AK097642.1 | - | 3 | 372 | ||
AK098452.1 | - | 3 | 475 | |||
AK294379.1 | - | 2 | 384 | |||
AK303005.1 | - | 3 | 501 | |||
AK309964.1 | - | 2 | 417 | |||
BC121161.1 | - | 2 | 311 | |||
BC121162.1 | - | 2 | 311 | |||
BC130011.1 | - | 2 | 311 | |||
BC153823.1 | - | 2 | 311 | |||
U56816.1 | - | 2 | 327 |