Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004693.2 | NP_004684.2 | 7, 8 | 1410 | ||
GenBank mRNA | AK225962.1 | - | 7, 8 | 1410 | ||
AK298960.1 | - | 7, 8 | 1609 | |||
Y17282.1 | - | 7, 8 | 1406 |