Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK223393.1 | - | 4 | 719 | ||
AK301154.1 | - | 4 | 742 | |||
AK303367.1 | - | 4 | 543 | |||
AK310676.1 | - | 4 | 581 | |||
AK313614.1 | - | 4 | 748 | |||
BC033583.1 | - | 4 | 703 | 5227138 | ||
CR996443.1 | - | 4 | 803 | |||
M14362.1 | - | 4 | 699 | |||
M16336.1 | - | 4 | 713 | |||
M16445.1 | - | 4 | 696 |