Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301645.1 | NP_001288574.1 | 2, 3 | 601 | ||
NM_004744.4 | NP_004735.2 | 2, 3 | 795 | |||
GenBank mRNA | AF071510.1 | - | 2, 3 | 634 | ||
AK292598.1 | - | 2, 3 | 795 | |||
AY546085.1 | - | 1, 2 | 869 | |||
AY546086.1 | - | 2, 3 | 766 | |||
BC031053.1 | - | 2, 3 | 604 | 5272486 |