Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB33A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004794.2 | NP_004785.1 | 2 | 756 | ||
XM_017029963.1 | XP_016885452.1 | 3 | 445 | |||
GenBank mRNA | AF498959.1 | - | 2 | 492 | ||
AK094927.1 | - | 3 | 1434 | |||
BC001157.1 | - | 2 | 756 | 3510745 | ||
BC009996.2 | - | 2 | 756 | 4129324 | ||
BT007038.1 | - | 2 | 492 | |||
D14889.1 | - | 2 | 929 |